Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP10 Antibody, Novus Biologicals™
SDP

Catalog No. NB11040545 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11040545 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11040545 Supplier Novus Biologicals Supplier No. NB11040545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

USP10 Polyclonal specifically detects USP10 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen USP10
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:100, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_005144.1
Gene Alias Deubiquitinating enzyme 10, EC 3.4.19.12, KIAA0190MGC2621, ubiquitin carboxyl-terminal hydrolase 10, ubiquitin specific peptidase 10, ubiquitin specific protease 10, ubiquitin thioesterase 10, Ubiquitin thiolesterase 10, Ubiquitin-specific-processing protease 10, UBPO
Gene Symbols USP10
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human Ubiquitin Specific Peptidase 10 using the numbering given in entry NP_005144.1 (GeneID 9100).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9100
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.