Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP10 Antibody (OTI2E1), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N274798P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274798P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274798P594 Supplier Novus Biologicals Supplier No. NBP274798PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP10 Monoclonal specifically detects USP10 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry, Knockout Validated.

Specifications

Antigen USP10
Applications Flow Cytometry, Immunoassay
Classification Monoclonal
Clone OTI2E1
Conjugate PE/ATTO 594
Dilution Flow Cytometry, Knockout Validated
Formulation PBS with 0.05% Sodium Azide
Gene Alias Deubiquitinating enzyme 10, EC 3.4.19.12, KIAA0190MGC2621, ubiquitin carboxyl-terminal hydrolase 10, ubiquitin specific peptidase 10, ubiquitin specific protease 10, ubiquitin thioesterase 10, Ubiquitin thiolesterase 10, Ubiquitin-specific-processing protease 10, UBPO
Gene Symbols USP10
Host Species Mouse
Immunogen Full length recombinant protein of human USP10 (NP_005144) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9100.0
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.