Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP11 Rabbit anti-Human, Mouse, Rat, Clone: 8I1K10, Novus Biologicals™
SDP

Catalog No. NB166650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166650 20 μg
NB166649 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166650 Supplier Novus Biologicals Supplier No. NBP31577720UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

USP11 Monoclonal antibody specifically detects USP11 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen USP11
Applications Western Blot
Classification Monoclonal
Clone 8I1K10
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Deubiquitinating enzyme 11, EC 3.1.2.15, EC 3.4.19.12, ubiquitin carboxyl-terminal hydrolase 11, ubiquitin specific peptidase 11, ubiquitin specific protease 11, ubiquitin thioesterase 11, Ubiquitin thiolesterase 11, ubiquitin-specific processing protease 11, Ubiquitin-specific-processing protease 11, UHX1ubiquitin carboxyl-terminal hydrolase, X-linked
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 800-900 of human USP11 (P51784).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Direct Reversal of DNA Damage
Primary or Secondary Primary
Gene ID (Entrez) 8237
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.