Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP12 Antibody (OTI1E3), Biotin, Novus Biologicals™
SDP

Catalog No. NBP274801B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274801B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274801B Supplier Novus Biologicals Supplier No. NBP274801B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP12 Monoclonal specifically detects USP12 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen USP12
Applications Western Blot
Classification Monoclonal
Clone OTI1E3
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias Deubiquitinating enzyme 12, EC 3.1.2.15, EC 3.4.19.12, UBH1ubiquitin specific protease 12, ubiquitin carboxyl-terminal hydrolase 12, ubiquitin specific peptidase 12, ubiquitin specific protease 12 like 1, ubiquitin thioesterase 12, Ubiquitin thiolesterase 12, Ubiquitin-hydrolyzing enzyme 1, Ubiquitin-specific-processing protease 12, USP12L1
Gene Symbols USP12
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 61-251 of human USP12 (NP_872294) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 219333.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.