Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP13 Antibody (OTI4G11), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP274802C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274802C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274802C Supplier Novus Biologicals Supplier No. NBP274802C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP13 Monoclonal specifically detects USP13 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen USP13
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4G11
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Deubiquitinating enzyme 13, EC 3.1.2.15, EC 3.4.19.12, Isopeptidase T-3, ISOT-3, ISOT3IsoT-3, ubiquitin carboxyl-terminal hydrolase 13, ubiquitin specific peptidase 13 (isopeptidase T-3), ubiquitin specific protease 13 (isopeptidase T-3), ubiquitin thioesterase 13, Ubiquitin thiolesterase 13, Ubiquitin-specific-processing protease 13
Gene Symbols USP13
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human USP13 expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 8975.0
Target Species Human, Mouse, Rat, Canine, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.