Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP14 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220825 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220825 Supplier Novus Biologicals Supplier No. NBP220825
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP14 Polyclonal specifically detects USP14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen USP14
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P54578
Gene Alias Deubiquitinating enzyme 14, EC 3.1.2.15, EC 3.4.19.12, TGT, TGTubiquitin carboxyl-terminal hydrolase 14, tRNA-guanine transglycosylase, 60-kD subunit, ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase), ubiquitin specific protease 14 (tRNA-guanine transglycosylase), ubiquitin thioesterase 14, Ubiquitin thiolesterase 14, ubiquitin-specific processing protease 14, Ubiquitin-specific-processing protease 14
Gene Symbols USP14
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human USP14. The exact sequence is proprietary.
Molecular Weight of Antigen 56 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Turnover, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 9097
Test Specificity Human, Mouse, Rat. Rhesus Monkey (100%), Chimpanzee (100%).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.