Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP19 Antibody, Novus Biologicals™
SDP

Catalog No. NB10079828 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10079828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10079828 Supplier Novus Biologicals Supplier No. NB10079828
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP19 Polyclonal specifically detects USP19 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen USP19
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_006668.1
Gene Alias Deubiquitinating enzyme 19, EC 3.1.2.15, EC 3.4.19.12, KIAA0891, ubiquitin carboxyl-terminal hydrolase 19, ubiquitin specific peptidase 19, ubiquitin thioesterase 19, Ubiquitin thiolesterase 19, Ubiquitin-specific-processing protease 19, Zinc finger MYND domain-containing protein 9, ZMYND9ubiquitin specific protease 19
Gene Symbols USP19
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human ubiquitin specific peptidase 19 (zinc finger MYND domain-containing protein 9) using the numbering given in entry NP_006668.1 (GeneID 10869).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10869
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.