Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110712
Description
Immunogen sequence: SGFPYGVTNN CLSYLPINAY DQGVTLTQKL DSQSDLARDF SSLRTSDSYR IDPRNLGRSP MLARTRKELC TLQGLYQTAS CPEYLVDYLE NYGRKGSASQ VPSQAPPSRV PEIISPTYR.
Ubiquitin is a highly conserved protein involved in the regulation of intracellular protein breakdown, cell cycle regulation, and stress response, is released from degraded proteins by disassembly of the polyubiquitin chains. The disassembly process is mediated by ubiquitin-specific proteases (USPs).
Specifications
USP2 | |
Polyclonal | |
Unconjugated | |
USP2 | |
41 kDa ubiquitin-specific protease; B930035K21Rik; deubiquitinating enzyme 2; MNCb-0190; Ubiquitin carboxyl-terminal hydrolase 2; ubiquitin specific peptidase 2; ubiquitin specific protease 12; ubiquitin specific protease 2; ubiquitin specific protease 9; ubiquitin thioesterase 2; ubiquitin thiolesterase 2; ubiquitin-specific-processing protease 2; Ubiquitin-specific-processing protease testis; Ubp41; Ubp69; UBP-t; USP2; USP9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9099 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.30 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75604 | |
USP2 | |
Recombinant Protein corresonding to Human USP2. Recombinant protein control fragment (Product #RP-89225). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction