Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | USP2 |
---|---|
Concentration | 0.30 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5110712
|
Invitrogen™
PA5110712 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5110723
|
Invitrogen™
PA5110723 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: SGFPYGVTNN CLSYLPINAY DQGVTLTQKL DSQSDLARDF SSLRTSDSYR IDPRNLGRSP MLARTRKELC TLQGLYQTAS CPEYLVDYLE NYGRKGSASQ VPSQAPPSRV PEIISPTYR.
Ubiquitin is a highly conserved protein involved in the regulation of intracellular protein breakdown, cell cycle regulation, and stress response, is released from degraded proteins by disassembly of the polyubiquitin chains. The disassembly process is mediated by ubiquitin-specific proteases (USPs).Specifications
USP2 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
USP2 | |
41 kDa ubiquitin-specific protease; B930035K21Rik; deubiquitinating enzyme 2; MNCb-0190; Ubiquitin carboxyl-terminal hydrolase 2; ubiquitin specific peptidase 2; ubiquitin specific protease 12; ubiquitin specific protease 2; ubiquitin specific protease 9; ubiquitin thioesterase 2; ubiquitin thiolesterase 2; ubiquitin-specific-processing protease 2; Ubiquitin-specific-processing protease testis; Ubp41; Ubp69; UBP-t; USP2; USP9 | |
USP2 | |
Primary | |
Antigen affinity chromatography |
0.30 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75604 | |
9099 | |
Recombinant Protein corresonding to Human USP2. Recombinant protein control fragment (Product #RP-89225). | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title