Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP20 Antibody, Novus Biologicals™
SDP

Catalog No. NB176767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176767 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176767 Supplier Novus Biologicals Supplier No. NBP313080
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USP20 Polyclonal antibody specifically detects USP20 in Human samples. It is validated for Western Blot

Specifications

Antigen USP20
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias Deubiquitinating enzyme 20, EC 3.4.19.12, KIAA1003hVDU2, LSFR3A, pVHL-interacting deubiquitinating enzyme 2, ubiquitin carboxyl-terminal hydrolase 20, ubiquitin specific peptidase 20, ubiquitin specific protease 20, ubiquitin thioesterase 20, Ubiquitin thiolesterase 20, Ubiquitin-specific-processing protease 20, VDU2, VHL-interacting deubiquitinating enzyme 2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human USP20. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10868
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.