Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP20 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB291699 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291699 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291699 Supplier Novus Biologicals Supplier No. NB10059837PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP20 Polyclonal antibody specifically detects USP20 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen USP20
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias Deubiquitinating enzyme 20, EC 3.4.19.12, KIAA1003hVDU2, LSFR3A, pVHL-interacting deubiquitinating enzyme 2, ubiquitin carboxyl-terminal hydrolase 20, ubiquitin specific peptidase 20, ubiquitin specific protease 20, ubiquitin thioesterase 20, Ubiquitin thiolesterase 20, Ubiquitin-specific-processing protease 20, VDU2, VHL-interacting deubiquitinating enzyme 2
Host Species Rabbit
Immunogen Maps to a region between residue 863 and 913 of human ubiquitin specific peptidase 20 (pVHL-interacting deubiquitinating enzyme 2) using the numbering given in entry NP_006667.2 (GeneID 10868).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10868
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.