Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP21 Antibody, Novus Biologicals™
SDP

Catalog No. NB093707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB093707 Supplier Novus Biologicals Supplier No. NBP303947
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP21 Polyclonal specifically detects USP21 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen USP21
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Deubiquitinating enzyme 21, EC 3.1.2.15, EC 3.4.19.12, MGC3394, ubiquitin carboxyl-terminal hydrolase 21, ubiquitin specific peptidase 21, ubiquitin specific protease 21, ubiquitin thioesterase 21, Ubiquitin thiolesterase 21, ubiquitin-specific processing protease 21, ubiquitin-specific protease 16, Ubiquitin-specific-processing protease 21, USP16, USP23NEDD8-specific protease
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 400-500 of human USP21 (NP_001014443.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Biology, Neuroscience, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 27005
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.