Learn More
Description
Specifications
Specifications
| Antigen | USP22 |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Gel Shift |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/ml, Simple Western 1:25, Chromatin Immunoprecipitation reported in scientific literature (PMID 34155658), Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 24197134), Gel Super Shift Assays reported in scientific literature, Proximity Ligation Assay reported in scientific literature (PMID 33198416), Chromatin Immunoprecipitation (ChIP) |
| Formulation | PBS and 30% Glycerol with 0.05% Sodium Azide |
| Gene Accession No. | Q9UPT9, Q5DU02 |
| Gene Alias | Deubiquitinating enzyme 22, EC 3.1.2.15, EC 3.4.19.12, KIAA1063ubiquitin carboxyl-terminal hydrolase 22, KIAA1064, ubiquitin specific peptidase 22, ubiquitin specific peptidase 3-like, ubiquitin specific protease 22, ubiquitin thioesterase 22, Ubiquitin thiolesterase 22, ubiquitin-specific processing protease 22, Ubiquitin-specific-processing protease 22, USP3L |
| Gene Symbols | USP22 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
