Learn More
Description
Specifications
Specifications
| Antigen | USP24 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Deubiquitinating enzyme 24, EC 3.4.19.12, FLJ31309, KIAA1057ubiquitin carboxyl-terminal hydrolase 24, ubiquitin specific peptidase 24, ubiquitin specific protease 24, ubiquitin thioesterase 24, Ubiquitin thiolesterase 24, ubiquitin-specific processing protease 24, Ubiquitin-specific-processing protease 24 |
| Gene Symbols | USP24 |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 2410 and the C-terminus (residue 2460) of human Ubiquitin Specific Peptidase 24 using the numbering given in entry XP_371254.5 (GeneID 23358). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
