Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP24 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB292047 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292047 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292047 Supplier Novus Biologicals Supplier No. NB10040830JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP24 Polyclonal specifically detects USP24 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen USP24
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias Deubiquitinating enzyme 24, EC 3.4.19.12, FLJ31309, KIAA1057ubiquitin carboxyl-terminal hydrolase 24, ubiquitin specific peptidase 24, ubiquitin specific protease 24, ubiquitin thioesterase 24, Ubiquitin thiolesterase 24, ubiquitin-specific processing protease 24, Ubiquitin-specific-processing protease 24
Host Species Rabbit
Immunogen Maps to a region between residue 2410 and the C-terminus (residue 2460) of human Ubiquitin Specific Peptidase 24 using the numbering given in entry XP_371254.5 (GeneID 23358).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 23358
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.