Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP25 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB184209 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184209 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184209 Supplier Novus Biologicals Supplier No. NBP282051AF594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USP25 Polyclonal antibody specifically detects USP25 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen USP25
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias Deubiquitinating enzyme 25, EC 3.1.2.15, EC 3.4.19.12, ubiquitin carboxyl-terminal hydrolase 25, ubiquitin specific peptidase 25, ubiquitin specific protease USP2510USP21ubiquitin specific protease 25, ubiquitin thioesterase 25, Ubiquitin thiolesterase 25, ubiquitin-specific processing protease 25, Ubiquitin-specific-processing protease 25, USP on chromosome 21
Host Species Rabbit
Immunogen USP25 antibody was raised against a 17 amino acid peptide near the center of human USP25. The immunogen is located within amino acids 470 - 520 of USP25. Amino Acid Squence: IPSQTLPSTTEQQGALS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 29761
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.