Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP28 Antibody, Novus Biologicals™
SDP

Catalog No. NB10096911 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10096911 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10096911 Supplier Novus Biologicals Supplier No. NB10096911
Add to Cart
Add to Cart

Goat Polyclonal Antibody

USP28 Polyclonal specifically detects USP28 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen USP28
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/ml, Immunohistochemistry 2.5 - 3.8 ug/ml, Immunohistochemistry-Paraffin 2.5 - 3.8 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_065937.1
Gene Alias Deubiquitinating enzyme 28, EC 3.1.2.15, EC 3.4.19.12, KIAA1515ubiquitin carboxyl-terminal hydrolase 28, ubiquitin specific peptidase 28, ubiquitin specific protease 28, ubiquitin thioesterase 28, Ubiquitin thiolesterase 28, Ubiquitin-specific-processing protease 28
Gene Symbols USP28
Host Species Goat
Immunogen Peptide with sequence C-SVELKHYIQEDN corresponding to internal region according to NP_065937.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57646
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Cow and Dog.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.