Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544965
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA544965 100 μL
1 options

Catalog No. PIPA544965

Supplier: Invitrogen™ PA544965

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EDILKEDNPV PNKKYKTDSL KYIQSNRKNP SSLEDLEKDR DLKLGPSFNT Sequence homology: Human: 100%.

USP29 (Ubiquitin Specific Peptidase 29) is a Protein Coding gene. Among its related pathways are Ubiquitin-Proteasome Dependent Proteolysis. Gene Ontology (GO) annotations related to this gene include cysteine-type endopeptidase activity and thiol-dependent ubiquitin-specific protease activity. An important paralog of this gene is USP37. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP29
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene USP29
Gene Accession No. Q9HBJ7
Gene Alias ASL1/Usp29 fusion; deubiquitinating enzyme 29; HOM-TES-84/86; Ocat; ossification center-associated transcript; ubiquitin carboxyl-terminal hydrolase 29; ubiquitin specific peptidase 29; ubiquitin specific protease 29; ubiquitin thioesterase 29; ubiquitin thiolesterase 29; ubiquitin-specific processing protease; ubiquitin-specific-processing protease 29; Usp29
Gene Symbols USP29
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human USP29 (aa 181-230).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57663
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.