Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP32 Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PA549541
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549541 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549541 Supplier Thermo Scientific Supplier No. PA549541

Rabbit Polyclonal Antibody

Thiol-dependent hydrolysis of ester, thioester, amide, peptide and isopeptide bonds formed by the C-terminal Gly of ubiquitin (a 76-residue protein attached to proteins as an intracellular targeting signal).
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP32
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene USP32
Gene Accession No. Q8NFA0
Gene Alias Ubiquitin carboxyl-terminal hydrolase 32, 3.4.19.12, Deubiquitinating enzyme 32, Renal carcinoma antigen NY-REN-60, Ubiquitin thioesterase 32, Ubiquitin-specific-processing protease 32, USP32, USP10
Gene Symbols USP32
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 438-470 amino acids from human USP32.
Purification Method Antigen affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84669
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.