Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP34 Antibody, Novus Biologicals™
SDP

Catalog No. NB1002863 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002863 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002863 Supplier Novus Biologicals Supplier No. NB1002863
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

USP34 Polyclonal specifically detects USP34 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen USP34
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. CAE51938.1
Gene Alias Deubiquitinating enzyme 34, EC 3.1.2.15, EC 3.4.19.12, FLJ43910, KIAA0570MGC104459, KIAA0729ubiquitin carboxyl-terminal hydrolase 34, ubiquitin specific peptidase 34, ubiquitin specific protease 34, ubiquitin thioesterase 34, Ubiquitin thiolesterase 34, Ubiquitin-specific-processing protease 34
Gene Symbols USP34
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 3350 and the C-terminus (residue 3395) of human Ubiquitin Specific Protease 34 using the numbering given in entry CAE51938.1 (GeneID 9736).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9736
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, European domestic ferret, and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.