Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP37 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220833 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220833 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220833 Supplier Novus Biologicals Supplier No. NBP220833
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP37 Polyclonal specifically detects USP37 in Human samples. It is validated for Western Blot.

Specifications

Antigen USP37
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q86T82
Gene Alias Deubiquitinating enzyme 37, EC 3.1.2.15, EC 3.4.19.12, KIAA1594tmp_locus_50, MGC117261, ubiquitin carboxyl-terminal hydrolase 37, ubiquitin specific peptidase 37, ubiquitin specific protease 37, ubiquitin thioesterase 37, Ubiquitin thiolesterase 37, Ubiquitin-specific-processing protease 37
Gene Symbols USP37
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human USP37. The exact sequence is proprietary.
Molecular Weight of Antigen 110 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Turnover, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 57695
Test Specificity Human. Species reactivity based on sequence identity: Mouse (87%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.