Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP37 Antibody, Novus Biologicals™
SDP

Catalog No. NB11040696 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11040696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11040696 Supplier Novus Biologicals Supplier No. NB11040696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP37 Polyclonal specifically detects USP37 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen USP37
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_065986.1
Gene Alias Deubiquitinating enzyme 37, EC 3.1.2.15, EC 3.4.19.12, KIAA1594tmp_locus_50, MGC117261, ubiquitin carboxyl-terminal hydrolase 37, ubiquitin specific peptidase 37, ubiquitin specific protease 37, ubiquitin thioesterase 37, Ubiquitin thiolesterase 37, Ubiquitin-specific-processing protease 37
Gene Symbols USP37
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 125 and 175 of human Ubiquitin Specific Peptidase 37 using the numbering given in entry NP_065986.1 (GeneID 57695).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57695
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.