Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP38 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB181194 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB181194 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB181194 Supplier Novus Biologicals Supplier No. NBP306481AF532
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USP38 Polyclonal antibody specifically detects USP38 in Human samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen USP38
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias Deubiquitinating enzyme 38, EC 3.4.19.12, HP43.8KDFLJ35970, KIAA1891ubiquitin specific protease 38, ubiquitin carboxyl-terminal hydrolase 38, ubiquitin specific peptidase 38, ubiquitin thioesterase 38, Ubiquitin thiolesterase 38, Ubiquitin-specific-processing protease 38
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human USP38 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84640
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.