Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP39 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP317961100UL
This item is not returnable.
View return policy
Description
USP39 Polyclonal antibody specifically detects USP39 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
USP39 | |
Polyclonal | |
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
CGI-21, HSPC332, Inactive ubiquitin-specific peptidase 39,65K, SAD1 homolog, SAD1MGC75069, small nuclear ribonucleoprotein 65kDa (U4/U6.U5), SnRNP assembly defective 1 homolog, SNRNP65, U4/U6.U5 tri-snRNP-associated 65 kDa protein, U4/U6.U5 tri-snRNP-associated protein 2, ubiquitin specific peptidase 39, ubiquitin specific protease 39 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: TIVNFPITNVDLREYLSEEVQAVHKNTTYDLIANIVHDGKPSEGSYRIHVLHHGTGKWYELQDLQVTDILPQMITLSEAYIQIWKRRDNDETN | |
100 μg | |
Cell Biology | |
10713 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction