Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP39 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$684.12
Specifications
| Antigen | USP39 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
USP39 Polyclonal antibody specifically detects USP39 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| USP39 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS, pH 7.2, 40% glycerol | |
| 10713 | |
| IgG | |
| Affinity purified |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CGI-21, HSPC332, Inactive ubiquitin-specific peptidase 39,65K, SAD1 homolog, SAD1MGC75069, small nuclear ribonucleoprotein 65kDa (U4/U6.U5), SnRNP assembly defective 1 homolog, SNRNP65, U4/U6.U5 tri-snRNP-associated 65 kDa protein, U4/U6.U5 tri-snRNP-associated protein 2, ubiquitin specific peptidase 39, ubiquitin specific protease 39 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: TIVNFPITNVDLREYLSEEVQAVHKNTTYDLIANIVHDGKPSEGSYRIHVLHHGTGKWYELQDLQVTDILPQMITLSEAYIQIWKRRDNDETN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title