Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP39 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734145 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
Catalog No. Quantity
50-173-4145 150 μL
1 options

Catalog No. 50-173-4145

Supplier: Proteintech Group Inc 238651AP150UL

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A novel gene product, hMAP126, interacts with p29 in the yeast two-hybrid assay. The subcellular distribution of hMAP126 was localized to the mitotic spindle and is phosphorylated by p34(cdc2) kinase. Human MAP126 is likely involved in the functional and dynamic regulation of mitotic spindles.

Specifications

Antigen USP39
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene USP39
Gene Accession No. Q53GS9
Gene Alias 65K, CGI 21, HSPC332, PRO2855, SAD1, SAD1 homolog, SNRNP65, USP39
Gene Symbols USP39
Host Species Rabbit
Immunogen USP39 Fusion Protein Ag20894
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10713
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.