Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP4 Antibody, Novus Biologicals™
SDP

Catalog No. NB1002868 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002868 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002868 Supplier Novus Biologicals Supplier No. NB1002868
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP4 Polyclonal specifically detects USP4 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen USP4
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2-5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. AAB72237.1
Gene Alias Deubiquitinating enzyme 4, EC 3.1.2.15, EC 3.4.19.12, MGC149848, MGC149849, ubiquitin carboxyl-terminal esterase 4, ubiquitin carboxyl-terminal hydrolase 4, ubiquitin specific peptidase 4 (proto-oncogene), ubiquitin specific protease 4 (proto-oncogene), ubiquitin thioesterase 4, Ubiquitin thiolesterase 4, ubiquitin-specific processing protease 4, Ubiquitin-specific-processing protease 4, Ubiquitous nuclear protein homolog, UNPH, UNPUnph
Gene Symbols USP4
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 225 and 275 of human Ubiquitin Specific Protease 4 using the numbering given in entry AAB72237.1 (GeneID 7375).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7375
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.