Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP42 Antibody, Novus Biologicals™
SDP

Catalog No. NB393914 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB393914 25 μL
NBP257538 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB393914 Supplier Novus Biologicals Supplier No. NBP25753825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP42 Polyclonal specifically detects USP42 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen USP42
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 μg/mL
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias Deubiquitinating enzyme 42, EC 3.1.2.15, EC 3.4.19.12, FLJ12697, ubiquitin carboxyl-terminal hydrolase 42, ubiquitin specific peptidase 42, ubiquitin specific protease 42, ubiquitin thioesterase 42, Ubiquitin thiolesterase 42, Ubiquitin-specific-processing protease 42
Gene Symbols USP42
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TIVDKASESSDPSAYQNQPGSSEAVSPGDMDAGSASWGAVSSLNDVSNHTLSLGPVPGAVVYSSSSVPDKSKPSPQKDQALGDGIAPPQKVL
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84132
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.