Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP43 Antibody, Novus Biologicals™
SDP

Catalog No. NB036184 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036184 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036184 Supplier Novus Biologicals Supplier No. NBP288562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP43 Polyclonal specifically detects USP43 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen USP43
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Formulation PBS, 2% Sucrose
Gene Alias Deubiquitinating enzyme 43, EC 3.1.2.15, EC 3.4.19.12, FLJ30626, ubiquitin carboxyl-terminal hydrolase 43, ubiquitin specific peptidase 43, ubiquitin specific protease 43, ubiquitin thioesterase 43, Ubiquitin thiolesterase 43, Ubiquitin-specific-processing protease 43
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of Human USP43
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124739
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.