Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP44 Antibody (OTI1D1), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP274811V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274811V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274811V Supplier Novus Biologicals Supplier No. NBP274811V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP44 Monoclonal specifically detects USP44 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen USP44
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1D1
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Deubiquitinating enzyme 44, DKFZp434D0127, EC 3.1.2.15, EC 3.4.19.12, FLJ14528, ubiquitin carboxyl-terminal hydrolase 44, ubiquitin specific peptidase 44, ubiquitin specific protease 44, ubiquitin thioesterase 44, Ubiquitin thiolesterase 44, Ubiquitin-specific-processing protease 44
Gene Symbols USP44
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 420-712 of human USP44 (NP_115523) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84101.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.