Learn More
Description
Specifications
Specifications
| Antigen | USP46 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | Deubiquitinating enzyme 46, EC 3.1.2.15, EC 3.4.19.12, FLJ11850, FLJ12552, FLJ14283, FLJ39393, ubiquitin carboxyl-terminal hydrolase 46, ubiquitin specific peptidase 46, ubiquitin specific protease 46, ubiquitin thioesterase 46, Ubiquitin thiolesterase 46, Ubiquitin-specific-processing protease 46 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant USP46 (NP_808229.1; Met1-Glu366). |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
