Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP47 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220836 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220836 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220836 Supplier Novus Biologicals Supplier No. NBP220836
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP47 Polyclonal specifically detects USP47 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen USP47
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.54mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q96K76
Gene Alias Deubiquitinating enzyme 47, DKFZp686C13257, EC 3.4.19.12, FLJ20727, Trf (TATA binding protein-related factor)-proximal homolog, Trf (TATA binding protein-related factor)-proximal homolog (Drosophila), TRFP, ubiquitin carboxyl-terminal hydrolase 47, ubiquitin specific peptidase 47, ubiquitin specific protease 47, ubiquitin thioesterase 47, Ubiquitin thiolesterase 47, Ubiquitin-specific-processing protease 47
Gene Symbols USP47
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human USP47. The exact sequence is proprietary.
Molecular Weight of Antigen 157 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55031
Test Specificity Human, Mouse. Species reactivity based on sequence identity: Chicken (94%), Rat (96%), Xenopus laevis (86%), Xenopus tropicalis (83%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.