Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP47 Antibody, Novus Biologicals™
SDP

Catalog No. NB10057487 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10057487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10057487 Supplier Novus Biologicals Supplier No. NB10057487
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP47 Polyclonal specifically detects USP47 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation.

Specifications

Antigen USP47
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunoprecipitation 2-5 mcg/mg, Immunohistochemistry-Paraffin 1:100-1:500
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_060414.3
Gene Alias Deubiquitinating enzyme 47, DKFZp686C13257, EC 3.4.19.12, FLJ20727, Trf (TATA binding protein-related factor)-proximal homolog, Trf (TATA binding protein-related factor)-proximal homolog (Drosophila), TRFP, ubiquitin carboxyl-terminal hydrolase 47, ubiquitin specific peptidase 47, ubiquitin specific protease 47, ubiquitin thioesterase 47, Ubiquitin thiolesterase 47, Ubiquitin-specific-processing protease 47
Gene Symbols USP47
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1237 and the C-terminus (residue 1287) of human ubiquitin specific peptidase 47 using the numbering given in entry NP_060414.3 (GeneID 55031).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55031
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.