Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP53 Antibody (OTI3C9), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP274812A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274812A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274812A70 Supplier Novus Biologicals Supplier No. NBP274812AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP53 Monoclonal specifically detects USP53 in Human samples. It is validated for Western Blot.

Specifications

Antigen USP53
Applications Western Blot
Classification Monoclonal
Clone OTI3C9
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DKFZp781E1417, inactive ubiquitin carboxyl-terminal hydrolase 53, Inactive ubiquitin-specific peptidase 53, KIAA1350, ubiquitin specific peptidase 53, ubiquitin specific protease 53, ubiquitin specific proteinase 53
Gene Symbols USP53
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 495-582 of human USP53 (NP_061923) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 54532.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.