Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP7 Antibody (OTI1F12), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N274814APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274814APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274814APC7 Supplier Novus Biologicals Supplier No. NBP274814APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP7 Monoclonal specifically detects USP7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen USP7
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1F12
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Deubiquitinating enzyme 7, EC 3.1.2.15, EC 3.4.19.12, HAUSPTEF1, Herpes virus-associated ubiquitin-specific protease, Herpesvirus-associated ubiquitin-specific protease, ubiquitin carboxyl-terminal hydrolase 7, ubiquitin specific peptidase 7 (herpes virus-associated), ubiquitin specific protease 7 (herpes virus-associated), Ubiquitin thioesterase 7, Ubiquitin-specific-processing protease 7
Gene Symbols USP7
Host Species Mouse
Immunogen Full length recombinant protein of human USP7 (NP_003461) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cell Biology, DNA Repair, Neuroscience, Stem Cell Markers, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7874.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.