Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP9x Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NBP148321
Description
USP9x Polyclonal specifically detects USP9x in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| USP9x | |
| Polyclonal | |
| Unconjugated | |
| PBS, 0.1% BSA, and 30% Glycerol with 0.05% Sodium Azide | |
| Deubiquitinating enzyme FAF-X, DFFRXubiquitin specific protease 9, X-linked (fat facets-like, Drosophila), Drosophila fat facets related, X-linked, EC 3.1.2.15, EC 3.4.19.12, FAF, FAM, Fat facets in mammals, fat facets protein related, X-linked, Fat facets protein-related, X-linked, hFAM, probable ubiquitin carboxyl-terminal hydrolase FAF-X, ubiquitin specific peptidase 9, X-linked, ubiquitin specific peptidase 9, X-linked (fat facets-like, Drosophila), ubiquitin thioesterase FAF-X, Ubiquitin thiolesterase FAF-X, ubiquitin-specific processing protease FAF-X, Ubiquitin-specific protease 9, X chromosome, Ubiquitin-specific-processing protease FAF-X, USP9, X chromosome (fat facets-like Drosophila) | |
| Rabbit | |
| 270 kDa | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Neuroscience | |
| 8239 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:25-1:200, Immunohistochemistry-Paraffin 1:50 | |
| Q93008, P70398 | |
| USP9X | |
| Genomic peptide made to an internal region of the human USP9x protein (within residues 1150-1300). [Swiss-Prot Q93008] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Immunogen sequence has 95% homology to rat and 80% homology to Zebrafish. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction