Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP9x Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 4 publications

$206.50 - $499.50

Specifications

Antigen USP9x
Concentration 0.2 mg/mL
Dilution Western Blot 1:1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:25-1:200, Immunohistochemistry-Paraffin 1:50
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP148321SS
SDP
View Documents
Novus Biologicals
NBP148321SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP148321
SDP
View Documents
Novus Biologicals
NBP148321
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

USP9x Polyclonal specifically detects USP9x in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

USP9x
Western Blot 1:1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:25-1:200, Immunohistochemistry-Paraffin 1:50
Polyclonal
Rabbit
Apoptosis, Autophagy, Cancer, Neuroscience
PBS, 0.1% BSA, and 30% Glycerol with 0.05% Sodium Azide
Deubiquitinating enzyme FAF-X, DFFRXubiquitin specific protease 9, X-linked (fat facets-like, Drosophila), Drosophila fat facets related, X-linked, EC 3.1.2.15, EC 3.4.19.12, FAF, FAM, Fat facets in mammals, fat facets protein related, X-linked, Fat facets protein-related, X-linked, hFAM, probable ubiquitin carboxyl-terminal hydrolase FAF-X, ubiquitin specific peptidase 9, X-linked, ubiquitin specific peptidase 9, X-linked (fat facets-like, Drosophila), ubiquitin thioesterase FAF-X, Ubiquitin thiolesterase FAF-X, ubiquitin-specific processing protease FAF-X, Ubiquitin-specific protease 9, X chromosome, Ubiquitin-specific-processing protease FAF-X, USP9, X chromosome (fat facets-like Drosophila)
USP9X
IgG
Affinity Purified
Human and mouse. Immunogen sequence has 95% homology to rat and 80% homology to Zebrafish.
0.2 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse
Q93008, P70398
8239
Genomic peptide made to an internal region of the human USP9x protein (within residues 1150-1300). [Swiss-Prot Q93008]
Primary
Store at -20C. Avoid freeze-thaw cycles.
270 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.