Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP9x Antibody (OTI2G7), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP274822C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274822C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274822C Supplier Novus Biologicals Supplier No. NBP274822C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP9x Monoclonal specifically detects USP9x in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen USP9x
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2G7
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Deubiquitinating enzyme FAF-X, DFFRXubiquitin specific protease 9, X-linked (fat facets-like, Drosophila), Drosophila fat facets related, X-linked, EC 3.1.2.15, EC 3.4.19.12, FAF, FAM, Fat facets in mammals, fat facets protein related, X-linked, Fat facets protein-related, X-linked, hFAM, probable ubiquitin carboxyl-terminal hydrolase FAF-X, ubiquitin specific peptidase 9, X-linked, ubiquitin specific peptidase 9, X-linked (fat facets-like, Drosophila), ubiquitin thioesterase FAF-X, Ubiquitin thiolesterase FAF-X, ubiquitin-specific processing protease FAF-X, Ubiquitin-specific protease 9, X chromosome, Ubiquitin-specific-processing protease FAF-X, USP9, X chromosome (fat facets-like Drosophila)
Gene Symbols USP9X
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 2246-2570 of human USP9x (NP_001034680) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Autophagy, Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8239.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.