Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP9x Antibody (OTI2G7), Novus Biologicals™
SDP

Catalog No. NBP203824 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP203824 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP203824 Supplier Novus Biologicals Supplier No. NBP203824
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP9x Monoclonal specifically detects USP9x in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen USP9x
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2G7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150
Gene Accession No. Q93008
Gene Alias Deubiquitinating enzyme FAF-X, DFFRXubiquitin specific protease 9, X-linked (fat facets-like, Drosophila), Drosophila fat facets related, X-linked, EC 3.1.2.15, EC 3.4.19.12, FAF, FAM, Fat facets in mammals, fat facets protein related, X-linked, Fat facets protein-related, X-linked, hFAM, probable ubiquitin carboxyl-terminal hydrolase FAF-X, ubiquitin specific peptidase 9, X-linked, ubiquitin specific peptidase 9, X-linked (fat facets-like, Drosophila), ubiquitin thioesterase FAF-X, Ubiquitin thiolesterase FAF-X, ubiquitin-specific processing protease FAF-X, Ubiquitin-specific protease 9, X chromosome, Ubiquitin-specific-processing protease FAF-X, USP9, X chromosome (fat facets-like Drosophila)
Gene Symbols USP9X
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 2246-2570 of human USP9X (NP_001034680) produced in E.coli.
Molecular Weight of Antigen 292 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8239
Target Species Human, Mouse, Rat, Canine, Primate
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.