Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP9X Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501560155
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0155 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0155 Supplier Bethyl Laboratories, Inc Supplier No. A301350AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

USP9X is an ubiquitin-specific protease involved in the removal and recycling of ubiquitin from proteins degraded in proteasomes. As a deubiquitinating enzyme, USP9x has a critical function in tight junction biogenesis by controlling the levels of the exchange factor for Arf6 (EFA6), a protein shown to facilitate tight junction formation during a narrow temporal window preceding the establishment of cell polarity (1). USP9X is also located on the X-chromosome but escapes X-inactivation, and defects in USP9X may cause gonadal degeneration in Turner syndrome.

Specifications

Antigen USP9X
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene USP9X
Gene Accession No. Q93008
Gene Alias DFFRX, FAF, FAM, Drosophila fat facets related, X-linked, deubiquitinating enzyme FAF-X, fat facets in mammals, fat facets protein related, X-linked, fat facets protein-related, X-linked, hFAM, probable ubiquitin carboxyl-terminal hydrolase FAF-X, ubiquitin specific protease 9, X chromosome (fat facets-like Drosophila), ubiquitin thioesterase FAF-X, ubiquitin thiolesterase FAF-X, ubiquitin-specific processing protease FAF-X, ubiquitin-specific protease 9, X chromosome, ubiquitin-specific-process
Gene Symbols USP9X
Host Species Rabbit
Immunogen Between 625 and 675
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8239
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.