Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP9Y Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179749
Description
USP9Y Polyclonal specifically detects USP9Y in Human samples. It is validated for Western Blot.Specifications
| USP9Y | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Deubiquitinating enzyme FAF-Y, DFFRYFLJ33043, EC 3.1.2.15, EC 3.4.19.12, Fat facets protein-related, Y-linked, probable ubiquitin carboxyl-terminal hydrolase FAF-Y, ubiquitin specific peptidase 9, Y-linked, ubiquitin specific peptidase 9, Y-linked (fat facets-like, Drosophila), ubiquitin thioesterase FAF-Y, Ubiquitin thiolesterase FAF-Y, ubiquitin-specific processing protease FAF-Y, Ubiquitin-specific protease 9, Y chromosome, Ubiquitin-specific-processing protease FAF-Y | |
| Rabbit | |
| 291 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Dog: 86%; Pig: 86%; Rat: 86%; Horse: 86%; Mouse: 86%; Bovine: 86%; Rabbit: 86%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_004645 | |
| USP9Y | |
| Synthetic peptide directed towards the C terminal of human USP9YThe immunogen for this antibody is USP9Y. Peptide sequence PHSPASQYQQNNHVHGQPYTGPAAHHLNNPQKTGQRTQENYEGNEEVSSP. | |
| Affinity purified | |
| RUO | |
| 8287 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction