Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP9Y Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167241 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167241 100 μg
NB167242 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167241 Supplier Novus Biologicals Supplier No. NBP316073100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USP9Y Polyclonal antibody specifically detects USP9Y in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen USP9Y
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 50% glycerol, pH7.3
Gene Alias Deubiquitinating enzyme FAF-Y, DFFRYFLJ33043, EC 3.1.2.15, EC 3.4.19.12, Fat facets protein-related, Y-linked, probable ubiquitin carboxyl-terminal hydrolase FAF-Y, ubiquitin specific peptidase 9, Y-linked, ubiquitin specific peptidase 9, Y-linked (fat facets-like, Drosophila), ubiquitin thioesterase FAF-Y, Ubiquitin thiolesterase FAF-Y, ubiquitin-specific processing protease FAF-Y, Ubiquitin-specific protease 9, Y chromosome, Ubiquitin-specific-processing protease FAF-Y
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 2456-2555 of human USP9Y (NP_004645.2).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8287
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.