Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UST Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen UST
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP160042
SDP
View Documents
Novus Biologicals
NBP160042
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

UST Polyclonal specifically detects UST in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

UST
Polyclonal
Purified
RUO
2OST, dermatan/chondroitin sulfate 2-sulfotransferase, DS2ST, EC 2.8.2.-, uronyl 2-sulfotransferase, uronyl-2-sulfotransferase
UST
IgG
Protein A purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Q9Y2C2
10090
Synthetic peptide directed towards the C terminal of human UST (NP_005706). Peptide sequence YFKGVLSIYKDPEHRKLGNMTVTVKKTVPSPEAVQILYQRMRYEYEFYHY.
Primary
48 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.