Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UT2/SLC14A2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP144375 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144375 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144375 Supplier Novus Biologicals Supplier No. NBP144375
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UT2/SLC14A2 Polyclonal specifically detects UT2/SLC14A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen UT2/SLC14A2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:25, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunohistochemistry-Paraffin
Formulation PBS and 50% Glycerol with 0.09% Sodium Azide
Gene Accession No. Q62668
Gene Alias FLJ16167, HUT2UT-A2, hUT-A6, MGC119566, solute carrier family 14 (urea transporter), member 2, Solute carrier family 14 member 2, urea transporter 2, Urea transporter, kidney, urea transporter-2, UT2MGC119567, UTA, UTR
Gene Symbols SLC14A2
Host Species Rabbit
Immunogen Produced against a synthetic peptide mapped to the C-terminal tail (amino acids 911-929) of rat UT-A1 (antibody designation L194)
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8170
Test Specificity Detects approx 97 and 127kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.