Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uteroglobin/SCGB1A1 Antibody (122), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB039807 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB039807 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB039807 Supplier Novus Biologicals Supplier No. NBP290517AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Uteroglobin/SCGB1A1 Monoclonal specifically detects Uteroglobin/SCGB1A1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Uteroglobin/SCGB1A1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 122
Conjugate Alexa Fluor 750
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias blastokinin, CC10Urine protein 1, CC16, CCSPCCPBP, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGBUP-1, UP1, Urinary protein 1, uteroglobin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Uteroglobin/SCGB1A1 (Q06318; Met1-Phe96).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 7356
Test Specificity No cross-reactivity in ELISA with: Human SCGB1A1
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.