Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uteroglobin/SCGB1A1 Antibody (21B2), Novus Biologicals™
SDP

Catalog No. NB298307 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298307 100 μL
NB298306 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298307 Supplier Novus Biologicals Supplier No. NBP326335100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Uteroglobin/SCGB1A1 Monoclonal antibody specifically detects Uteroglobin/SCGB1A1 in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen Uteroglobin/SCGB1A1
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 21B2
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias blastokinin, CC10Urine protein 1, CC16, CCSPCCPBP, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGBUP-1, UP1, Urinary protein 1, uteroglobin
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Uteroglobin/SCGB1A1 [UniProt P11684]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 7356
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.