Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uteroglobin/SCGB1A1 Antibody (3A8B8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237439
Description
Uteroglobin/SCGB1A1 Monoclonal specifically detects Uteroglobin/SCGB1A1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
Uteroglobin/SCGB1A1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
blastokinin, CC10Urine protein 1, CC16, CCSPCCPBP, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGBUP-1, UP1, Urinary protein 1, uteroglobin | |
Mouse | |
10 kDa | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research | |
7356 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
3A8B8 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
SCGB1A1 | |
Purified recombinant fragment of human SCGB1A1 (AA: 26-91) expressed in E. Coli. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction