Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uteroglobin/SCGB1A1 Antibody (3A8B8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$562.50
Specifications
Antigen | Uteroglobin/SCGB1A1 |
---|---|
Clone | 3A8B8 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
Description
Uteroglobin/SCGB1A1 Monoclonal specifically detects Uteroglobin/SCGB1A1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
Uteroglobin/SCGB1A1 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
Unconjugated | |
RUO | |
Human | |
blastokinin, CC10Urine protein 1, CC16, CCSPCCPBP, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGBUP-1, UP1, Urinary protein 1, uteroglobin | |
SCGB1A1 | |
IgG1 | |
Affinity Purified |
3A8B8 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Mouse | |
Biologically Active Proteins, Cytokine Research | |
PBS with 0.05% Sodium Azide | |
7356 | |
Purified recombinant fragment of human SCGB1A1 (AA: 26-91) expressed in E. Coli. | |
Primary | |
10 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title