Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UTF1 Mouse anti-Human, Clone: E0-F4, Novus Biologicals™
SDP

Catalog No. NBP266773 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266773 Supplier Novus Biologicals Supplier No. NBP266773

Mouse Monoclonal Antibody

UTF1 Monoclonal specifically detects UTF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen UTF1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone E0-F4
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:1000
Formulation PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias undifferentiated embryonic cell transcription factor 1
Gene Symbols UTF1
Host Species Mouse
Immunogen Synthetic peptide within Human UTF1 aa 51-100/ 341. (SwissProt: Q5T230 Human)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8433
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.