Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UTP11L Antibody (OTI5F3), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP274823A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274823A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274823A30 Supplier Novus Biologicals Supplier No. NBP274823AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UTP11L Monoclonal specifically detects UTP11L in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen UTP11L
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5F3
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CGI94, CGI-94, probable U3 small nucleolar RNA-associated protein 11, U3 snoRNA-associated protein 11, UTP11-like protein, UTP11-like, U3 small nucleolar ribonucleoprotein, (yeast)
Gene Symbols UTP11L
Host Species Mouse
Immunogen Full length recombinant protein of human UTP11L (NP_057121) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 51118
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.